6,6'-Dibromo-1,1'-bis(2-exylhexyl)isoindigo 97%

Supplier: Ambeed
Warning

A334264-1G A334264-250MG
77167-454EA 156 USD
77167-454 77167-456
6,6'-Dibromo-1,1'-bis(2-exylhexyl)isoindigo 97%
6,6'-Dibromo-1,1'-bis(2-exylhexyl)isoindigo

Precaución: for Research Use Only

Formula: C₃₂H₄₀Br₂N₂O₂
MDL Number: MFCD28125575
CAS Number: 1147124-23-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR