1-((1,3-Dioxolan-2-yl)methyl)-1H-pyrazole-4-boronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 1h-pyrazole, 1-(1,3-dioxolan-2-ylmethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-, 1-(1,3-Dioxolan-2-ylmethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

A333808-100MG A333808-1G A333808-250MG A333808-5G
77195-036EA 49 USD
77195-036 77195-038 77195-040 77195-042
1-((1,3-Dioxolan-2-yl)methyl)-1H-pyrazole-4-boronic acid pinacol ester 98%
1-((1,3-Dioxolan-2-yl)methyl)-1H-pyrazole-4-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₂₁BN₂O₄
MDL Number: MFCD08272028
CAS Number: 864754-17-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR