1,4,7-Triazacyclononane 96%

Supplier: Ambeed
Danger

Synonyms: 1,4,7-Triazonane, TACN

A331209-5G A331209-1G
77195-010EA 488.22 USD
77195-010 77550-936
1,4,7-Triazacyclononane 96%
1,4,7-Triazacyclononane

Precaución: for Research Use Only

Formula: C₆H₁₅N₃
MW: 129.2 g/mol
MDL Number: MFCD00012358
CAS Number: 4730-54-5
UN: 3259
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR