tert-Butyl-N-(Benzyloxy)carbamate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl benzyloxycarbamate, N-(Benzyloxy)carbamic Acid tert-Butyl Ester
A331053-10G
A331053-1G
A331053-25G
A331053-5G
A331053-100G
77167-432EA
32.96
USD
77167-432
77167-434
77167-436
77167-438
77683-989
tert-Butyl-N-(Benzyloxy)carbamate 97%
tert-Butyl-N-(Benzyloxy)carbamate
Precaución: For research use only.
Formula:
C₁₂H₁₇NO₃ MW: 223.27 g/mol |
MDL Number:
MFCD00191873 CAS Number: 79722-21-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...