(2-Cyanopyrimidin-5-yl)boronic acid 98%

Supplier: Ambeed
Warning

A330774-1G A330774-250MG A330774-100MG
77167-422EA 395.52 USD
77167-422 77167-424 77683-988
(2-Cyanopyrimidin-5-yl)boronic acid 98%
(2-Cyanopyrimidin-5-yl)boronic acid

Precaución: For research use only.

Formula: C₅H₄BN₃O₂
MW: 148.91 g/mol
MDL Number: MFCD10697511
CAS Number: 1164100-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR