4-Chloro-6-methyl-2-(methylthio)pyrimidine-5-carbonitrile 97%
Supplier: Ambeed
|
Warning
|
A330383-100MG
A330383-250MG
A330383-1G
A330383-5G
77167-416EA
32.96
USD
77167-416
77167-418
77612-898
77683-986
4-Chloro-6-methyl-2-(methylthio)pyrimidine-5-carbonitrile 97%
4-Chloro-6-methyl-2-(methylthio)pyrimidine-5-carbonitrile
Precaución: For research use only.
Formula:
C₇H₆ClN₃S MW: 199.661 g/mol |
MDL Number:
MFCD28134480 CAS Number: 1208170-17-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...