2-Chloro-1-isopropyl-1H-benzo[d]imidazole 95%

Supplier: Ambeed
Warning

A330254-100MG A330254-1G A330254-250MG
77167-410EA 29.87 USD
77167-410 77167-412 77167-414
2-Chloro-1-isopropyl-1H-benzo[d]imidazole 95%
2-Chloro-1-isopropyl-1H-benzo[d]imidazole

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClN₂
MW: 194.66 g/mol
MDL Number: MFCD00456409
CAS Number: 3705-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR