(R)-2-Acetamido-3-(naphtalen-2-YL)propanoic acid 97%

Supplier: Ambeed
Warning

Synonyms: Ac-D-2-Nal-OH, N-Acetyl-3-(2-naphthyl)-D-alanine

A329813-1G A329813-5G
77167-406EA 54 USD
77167-406 77167-408
(R)-2-Acetamido-3-(naphtalen-2-YL)propanoic acid 97%
(R)-2-Acetamido-3-(naphtalen-2-YL)propanoic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₅NO₃
MW: 257.289 g/mol
MDL Number: MFCD00079678
CAS Number: 37440-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR