Methyl 6-(benzyloxy)-4-chloroquinoline-2-carboxylate 97%

Supplier: Ambeed
Warning

A329349-100MG A329349-10G A329349-1G A329349-250MG
77194-974EA 38 USD
77194-974 77194-976 77194-978 77194-980
Methyl 6-(benzyloxy)-4-chloroquinoline-2-carboxylate 97%
Methyl 6-(benzyloxy)-4-chloroquinoline-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₈H₁₄ClNO₃
MDL Number: MFCD08458391
CAS Number: 905807-66-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR