Methyl 3-fluoroisonicotinate 96%

Supplier: Ambeed
Warning

A328883-100MG A328883-1G A328883-250MG A328883-5G
77167-388EA 5 USD
77167-388 77167-390 77167-392 77167-394
Methyl 3-fluoroisonicotinate 96%
Methyl 3-fluoroisonicotinate

Precaución: for Research Use Only

Formula: C₇H₆FNO₂
MDL Number: MFCD11045048
CAS Number: 876919-08-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR