1,2,4,5-Tetraiodobenzene 95%

Supplier: Ambeed
Danger

A328800-100MG A328800-1G A328800-250MG
77194-964EA 28.84 USD
77194-964 77194-966 77194-968
1,2,4,5-Tetraiodobenzene 95%
1,2,4,5-Tetraiodobenzene

Precaución: for Research Use Only

Formula: C₆H₂I₄
MDL Number: MFCD02684763
CAS Number: 636-31-7
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR