2-Phenoxybenzyl chloride 98%

Supplier: Ambeed
Danger

A328793-25G A328793-5G
77194-960EA 142 USD
77194-960 77194-962
2-Phenoxybenzyl chloride 98%
2-Phenoxybenzyl chloride

Precaución: for Research Use Only

Formula: C₁₃H₁₁ClO
MDL Number: MFCD00449749
CAS Number: 5888-53-9
UN: 1760
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR