1-(3-Methoxyphenyl)-2-thiourea 95%

Supplier: Ambeed
Danger

Synonyms: 3-Methoxyphenylthiourea, 1-(3-Methoxyphenyl)thiourea, N-(3-Methoxyphenyl)thiourea

A328266-1G A328266-5G
77194-944EA 21.63 USD
77194-944 77194-946
1-(3-Methoxyphenyl)-2-thiourea 95%
1-(3-Methoxyphenyl)-2-thiourea

Precaución: for Research Use Only

Formula: (C₆H₄(OCH₃))NC(S)NH₂
MW: 182.25 g/mol
MDL Number: MFCD00060446
CAS Number: 37014-08-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR