4-[(Piperidin-4-yl)oxy]piperidine-1-carboxylic acid tert-butyl ester 98%

Supplier: Ambeed
Warning

A328110-100MG A328110-1G A328110-250MG A328110-5G
77167-378EA 58.71 USD
77167-378 77167-380 77167-382 77683-980
4-[(Piperidin-4-yl)oxy]piperidine-1-carboxylic acid tert-butyl ester 98%
4-[(Piperidin-4-yl)oxy]piperidine-1-carboxylic acid tert-butyl ester

Precaución: For research use only.

Formula: C₁₅H₂₈N₂O₃
MW: 284.39 g/mol
MDL Number: MFCD09261176
CAS Number: 845305-83-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR