4-(4-Bromo-3-(1,3-dioxolan-2-yl)phenoxy)benzonitrile 95%

Supplier: Ambeed
Warning

A328066-1G A328066-250MG A328066-5G
77167-372EA 25 USD
77167-372 77167-374 77167-376
4-(4-Bromo-3-(1,3-dioxolan-2-yl)phenoxy)benzonitrile 95%
4-(4-Bromo-3-(1,3-dioxolan-2-yl)phenoxy)benzonitrile

Precaución: for Research Use Only

Formula: C₁₆H₁₂BrNO₃
MDL Number: MFCD28134561
CAS Number: 1217366-74-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR