5-Amino-2-chloronicotinic acid 98%

Supplier: Ambeed
Warning

A327196-100MG A327196-1G A327196-250MG A327196-5G
77167-364EA 8 USD
77167-364 77167-366 77167-368 77892-674
5-Amino-2-chloronicotinic acid 98%
5-Amino-2-chloronicotinic acid

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O₂
MDL Number: MFCD06658302
CAS Number: 42959-39-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR