4-(Morpholinosulfonyl)phenol 97%

Supplier: Ambeed
Warning

A327017-100MG A327017-1G A327017-250MG
77194-924EA 38 USD
77194-924 77194-926 77194-928
4-(Morpholinosulfonyl)phenol 97%
4-(Morpholinosulfonyl)phenol

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₄S
MDL Number: MFCD03147214
CAS Number: 3077-65-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR