(10E,12Z)-Octadeca-10,12-dienoic acid 80% (mixture of isomers)

Supplier: Ambeed
Warning

A326629-25G A326629-500G A326629-100G
77194-916EA 7 USD
77194-916 77194-918 77746-637
(10E,12Z)-Octadeca-10,12-dienoic acid 80% (mixture of isomers)
(10E,12Z)-Octadeca-10,12-dienoic acid

Precaución: for Research Use Only

Formula: C₁₈H₃₂O₂
MDL Number: MFCD02259040
CAS Number: 2420-56-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR