3-Chlorobenzamidine hydrochloride 98%

Supplier: Ambeed
Warning

A326520-1G A326520-5G
77167-356EA 30 USD
77167-356 77167-358 77167-360
3-Chlorobenzamidine hydrochloride 98%
3-Chlorobenzamidine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₈Cl₂N₂
MDL Number: MFCD00276754
CAS Number: 24095-60-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR