(5-Chloro-2-methoxyphenyl)methanamine hydrochloride 95%

Supplier: Ambeed
Warning

A325599-100MG A325599-1G A325599-250MG
77167-346EA 16 USD
77167-346 77167-348 77167-350
(5-Chloro-2-methoxyphenyl)methanamine hydrochloride 95%
(5-Chloro-2-methoxyphenyl)methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₁Cl₂NO
MDL Number: MFCD06797811
CAS Number: 350480-55-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR