3-Bromo-4-chloro-5-methoxyaniline 95%

Supplier: Ambeed
Warning

A325465-1G A325465-250MG A325465-5G
77194-910EA 55 USD
77194-910 77194-912 77194-914
3-Bromo-4-chloro-5-methoxyaniline 95%
3-Bromo-4-chloro-5-methoxyaniline

Precaución: for Research Use Only

Formula: C₇H₇BrClNO
MDL Number: MFCD18089325
CAS Number: 940948-33-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR