1,1,1-Tris(4-hydroxyphenyl)ethane 98%

Supplier: Ambeed
Danger

Synonyms: 4,4',4''-(Ethane-1,1,1-triyl)triphenol

A324310-10G A324310-25G A324310-5G A324310-100G
77194-894EA 7.21 USD
77194-894 77194-896 77194-898 77571-918
1,1,1-Tris(4-hydroxyphenyl)ethane 98%
1,1,1-Tris(4-hydroxyphenyl)ethane

Precaución: for Research Use Only

Formula: C₂₀H₁₈O₃
MW: 306.36 g/mol
MDL Number: MFCD00012180
CAS Number: 27955-94-8
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR