3,7-Dibromoimidazo[1,2-a]pyridine 98%

Supplier: Ambeed
Danger

A322295-100MG A322295-1G A322295-250MG
77194-856EA 11 USD
77194-856 77194-858 77194-860
3,7-Dibromoimidazo[1,2-a]pyridine 98%
3,7-Dibromoimidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₇H₄Br₂N₂
MDL Number: MFCD22551923
CAS Number: 1263060-75-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR