9-Bromo-7H-benzo[c]carbazole 98%

Supplier: Ambeed
Warning

A322238-1G A322238-25G A322238-5G
77167-324EA 17.51 USD
77167-324 77167-326 77167-328
9-Bromo-7H-benzo[c]carbazole 98%
9-Bromo-7H-benzo[c]carbazole

Precaución: for Research Use Only

Formula: C₁₆H₁₀BrN
MW: 296.16 g/mol
MDL Number: MFCD29918859
CAS Number: 1357572-66-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR