5-Bromo-2-(piperazin-1yl)pyrimidine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-(1-piperazinyl)pyrimidine

A321963-100G A321963-1G A321963-25G A321963-5G
77194-848EA 714.82 USD
77194-848 77194-850 77194-852 77194-854
5-Bromo-2-(piperazin-1yl)pyrimidine 98%
5-Bromo-2-(piperazin-1yl)pyrimidine

Precaución: for Research Use Only

Formula: C₈H₁₁BrN₄
MW: 243.11 g/mol
MDL Number: MFCD01314280
CAS Number: 99931-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR