Tris(triphenylphosphine)rhodium(I) chloride ≥99.9% (trace metals basis)

Supplier: Ambeed
Warning

Synonyms: Wilkinson's catalyst, Chlorotris(triphenylphosphine)rhodium(I), Tris(triphenylphosphine)chlororhodium

A321287-100MG A321287-1G A321287-250MG A321287-5G
77194-832EA 16 USD
77194-832 77194-834 77194-836 77568-642
Tris(triphenylphosphine)rhodium(I) chloride ≥99.9% (trace metals basis)
Tris(triphenylphosphine)rhodium(I) chloride

Precaución: For research use only.

Formula: C₅₄H₄₅ClP₃Rh
MW: 925.23 g/mol
Melting Pt: 157
MDL Number: MFCD00010016
CAS Number: 14694-95-2
Merck Index: 12,02226

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR