4-(4-Methylpiperidin-1-yl)benzoic acid 97%

Supplier: Ambeed
Warning

A321167-1G A321167-250MG A321167-100MG
77194-828EA 106.09 USD
77194-828 77194-830 77746-621
4-(4-Methylpiperidin-1-yl)benzoic acid 97%
4-(4-Methylpiperidin-1-yl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO₂
MW: 219.27 g/mol
MDL Number: MFCD04113993
CAS Number: 97096-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR