Methyl-4-aminocyclohexanecarboxylate hydrochloride (cis and trans mixture) 97%

Supplier: Ambeed
Danger

Synonyms: 4-Amino-cyclohexanecarboxylic acid methyl ester hydrochloride, Methyl 4-aminocyclohexanecarboxylate hydrochloride, 4-Aminocyclohexanecarboxylic Acid Methyl Ester Hydrochloride

A320346-5G A320346-250MG A320346-1G A320346-25G
77167-272EA 271.92 USD
77167-272 77612-852 77683-954 77683-955
Methyl-4-aminocyclohexanecarboxylate hydrochloride (cis and trans mixture) 97%
Methyl-4-aminocyclohexanecarboxylate hydrochloride (cis and trans mixture)

Precaución: For research use only.

Formula: C₈H₁₅NO₂
MW: 193.67 g/mol
MDL Number: MFCD11870896
CAS Number: 100707-54-8
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR