Dimethyl 3,4-dihydroxythiophene-2,5-dicarboxylate, disodium salt 95+%

Supplier: Ambeed
Warning

A319149-100MG A319149-1G
77167-250EA 39.14 USD
77167-250 77167-252
Dimethyl 3,4-dihydroxythiophene-2,5-dicarboxylate, disodium salt 95+%
Dimethyl 3,4-dihydroxythiophene-2,5-dicarboxylate, disodium salt

Precaución: for Research Use Only

Formula: C₈H₆Na₂O₆S
MW: 276.17 g/mol
MDL Number: MFCD16621664
CAS Number: 108199-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR