3-Bromo-1,4-dimethyl-1H-pyrazole 98%

Supplier: Ambeed
Warning

A319015-100MG A319015-250MG A319015-1G
77167-246EA 105.06 USD
77167-246 77167-248 77550-888
3-Bromo-1,4-dimethyl-1H-pyrazole 98%
3-Bromo-1,4-dimethyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₅H₇BrN₂
MW: 175.02 g/mol
MDL Number: MFCD28099396
CAS Number: 13745-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR