CAY-10415 97%

Supplier: Ambeed
Warning

Synonyms: AC1MIM9Y, MSDC 0160, 2-(1-oxo-4,4-diphenyl-2,3,4a,5,7,7a-hexahydrothiopyrano[2,3-c]pyrrol-6-yl)-1-(4-pyrrolidin-1-ylphenyl)ethanone, Thiopyrano(2,3-c)pyrrole, octahydro-4,4-diphenyl-6-((2-(1-pyrrolidinyl)phenyl)acetyl)-, 1-oxide, (1-ɑ,4a-β,7a-β)-(+-)-, 5-(4-(2-(5-Ethylpyridin-2-yl)-2-oxoethoxy)benzyl)thiazolidine-2,4-dione, 5-[[4-[2-(5-ethylpyridin-2-yl)-2-oxoethoxy]phenyl]methyl]-1,3-thiazolidine-2,4-dione, HE257587

A318812-10MG A318812-1MG A318812-2MG
77194-816EA 79.31 USD
77194-816 77194-818 77194-820
CAY-10415 97%
CAY-10415

Precaución: for Research Use Only

Formula: C₁₉H₁₈N₂O₄S
MW: 370.423 g/mol
MDL Number: MFCD00916084
CAS Number: 146062-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR