2-Fluoro-5-(trifluoromethyl)isonicotinic acid 97%

Supplier: Ambeed
Warning

A318809-100MG A318809-250MG A318809-1G
77194-812EA 110.21 USD
77194-812 77194-814 77746-612
2-Fluoro-5-(trifluoromethyl)isonicotinic acid 97%
2-Fluoro-5-(trifluoromethyl)isonicotinic acid

Precaución: for Research Use Only

Formula: C₇H₃F₄NO₂
MW: 209.09 g/mol
MDL Number: MFCD16611302
CAS Number: 1227574-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR