1-(1H-Indazol-3-yl)ethanone 97%

Supplier: Ambeed
Warning

A318571-1G A318571-250MG A318571-5G
77194-806EA 66.95 USD
77194-806 77194-808 77194-810
1-(1H-Indazol-3-yl)ethanone 97%
1-(1H-Indazol-3-yl)ethanone

Precaución: for Research Use Only

Formula: C₉H₈N₂O
MW: 160.17 g/mol
MDL Number: MFCD08234921
CAS Number: 4498-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR