Tris[2-(4,6-difluorophenyl)pyridinato-C2,N]iridium(III) 98%

Supplier: Ambeed
Warning

A318078-100MG A318078-250MG A318078-1G
77194-802EA 69 USD
77194-802 77194-804 77746-611
Tris[2-(4,6-difluorophenyl)pyridinato-C2,N]iridium(III) 98%
Tris[2-(4,6-difluorophenyl)pyridinato-C2,N]iridium(III)

Precaución: for Research Use Only

Formula: C₃₃H₁₈F₆IrN₃
MDL Number: MFCD09842738
CAS Number: 387859-70-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR