7-Bromo-5-methylquinoxaline 97%

Supplier: Ambeed
Warning

A317901-100MG A317901-1G A317901-250MG
77194-794EA 18 USD
77194-794 77194-796 77194-798
7-Bromo-5-methylquinoxaline 97%
7-Bromo-5-methylquinoxaline

Precaución: for Research Use Only

Formula: C₉H₇BrN₂
MDL Number: MFCD16987767
CAS Number: 532934-95-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR