1-(3-Hydroxypropyl)-2-pyrrolidone 98%

Supplier: Ambeed
Warning

A317589-1G A317589-25G A317589-5G
77194-788EA 20 USD
77194-788 77194-790 77194-792
1-(3-Hydroxypropyl)-2-pyrrolidone 98%
1-(3-Hydroxypropyl)-2-pyrrolidone

Precaución: for Research Use Only

Formula: C₇H₁₃NO₂
MDL Number: MFCD00059014
CAS Number: 62012-15-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR