9-Mesityl-10-methylacridin-10-ium tetrafluoroborate 97%

Supplier: Ambeed
Warning

A316965-100MG A316965-1G A316965-250MG A316965-5G
77194-772EA 27 USD
77194-772 77194-774 77194-776 77194-778
9-Mesityl-10-methylacridin-10-ium tetrafluoroborate 97%
9-Mesityl-10-methylacridin-10-ium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₂₃H₂₂BF₄N
MDL Number: MFCD28411690
CAS Number: 1442433-71-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR