5-(4-Bromophenyl)pyrimidine-4,6-diol 97%

Supplier: Ambeed
Warning

Synonyms: 5-(4-Bromophenyl)-6-hydroxypyrimidin-4(1H)-one

A316950-100G A316950-10G A316950-1G A316950-25G A316950-5G
77167-230EA 400.67 USD
77167-230 77167-232 77167-234 77167-236 77167-238
5-(4-Bromophenyl)pyrimidine-4,6-diol 97%
5-(4-Bromophenyl)pyrimidine-4,6-diol

Precaución: for Research Use Only

Formula: C₁₀H₇BrN₂O₂
MW: 267.079 g/mol
MDL Number: MFCD13151898
CAS Number: 706811-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR