(3-Methyl-4-(methylthio)phenyl)boronic acid 96%

Supplier: Ambeed
Warning

Synonyms: 3-Methyl-4-(methylthio)phenylboronic acid

A316264-100MG A316264-1G A316264-250MG
77194-758EA 37 USD
77194-758 77194-760 77194-762
(3-Methyl-4-(methylthio)phenyl)boronic acid 96%
(3-Methyl-4-(methylthio)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BO₂S
MDL Number: MFCD28147715
CAS Number: 221031-07-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR