1-Benzyl-3-(methylamino)pyrrolidine 97%

Supplier: Ambeed
Warning

Synonyms: 1-Benzyl-N-methylpyrrolidin-3-amine

A316171-250MG A316171-1G
77194-756EA 14.42 USD
77194-756 77746-606
1-Benzyl-3-(methylamino)pyrrolidine 97%
1-Benzyl-3-(methylamino)pyrrolidine

Precaución: for Research Use Only

Formula: C₁₂H₁₈N₂
MW: 190.29 g/mol
MDL Number: MFCD00191308
CAS Number: 96568-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR