(S)-tert-Butyl 2-acetylpyrrolidine-1-carboxylate 97%

Supplier: Ambeed
Warning

A316077-100MG A316077-1G A316077-250MG
77167-224EA 28.84 USD
77167-224 77167-226 77167-228
(S)-tert-Butyl 2-acetylpyrrolidine-1-carboxylate 97%
(S)-tert-Butyl 2-acetylpyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₃
MW: 213.27 g/mol
MDL Number: MFCD11975893
CAS Number: 91550-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR