Chloro(2-dicyclohexylphosphino-2',6'-dimethoxy-1,1'-biphenyl)(2'-amino-1,1'-biphenyl-2-yl)palladium(II) 98%
Supplier: Ambeed
|
Warning
|
A315893-100MG
A315893-1G
A315893-250MG
A315893-5G
A315893-25G
77167-216EA
11
USD
77167-216
77167-218
77167-220
77167-222
77550-884
Chloro(2-dicyclohexylphosphino-2',6'-dimethoxy-1,1'-biphenyl)(2'-amino-1,1'-biphenyl-2-yl)palladium(II) 98%
Chloro(2-dicyclohexylphosphino-2',6'-dimethoxy-1,1'-biphenyl)(2'-amino-1,1'-biphenyl-2-yl)palladium(II)
Precaución: for Research Use Only
Formula:
C₃₈H₄₅ClNO₂PPd |
MDL Number:
MFCD20264897 CAS Number: 1375325-64-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...