Methyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 1H-pyrazolo[3,4-b]pyridine-5-carboxylate

A315228-100MG A315228-1G
77167-198EA 86.52 USD
77167-198 77167-200
Methyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylate 98%
Methyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylate

Precaución: for Research Use Only

Formula: C₈H₇N₃O₂
MW: 177.16 g/mol
MDL Number: MFCD11977789
CAS Number: 1196156-42-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR