4-Bromo-2-(tert-butyl)benzonitrile 95%
Supplier: Ambeed
|
Warning
|
A314683-100MG
A314683-250MG
77167-196EA
6
USD
77167-196
77892-645
4-Bromo-2-(tert-butyl)benzonitrile 95%
4-Bromo-2-(tert-butyl)benzonitrile
Precaución: for Research Use Only
Formula:
C₁₁H₁₂BrN |
MDL Number:
MFCD28963501 CAS Number: 1369819-64-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...