3-Phenyl-9H-carbazole 98%

Supplier: Ambeed
Warning

A314615-10G A314615-250MG A314615-5G A314615-1G
77194-726EA 32.96 USD
77194-726 77194-728 77194-730 77746-602
3-Phenyl-9H-carbazole 98%
3-Phenyl-9H-carbazole

Precaución: for Research Use Only

Formula: C₁₈H₁₃N
MW: 243.31 g/mol
MDL Number: MFCD22052094
CAS Number: 103012-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR