1-(2,3-Di(thiophen-2-yl)quinoxalin-6-yl)-3-(2-methoxyethyl)urea 96%

Supplier: Ambeed
Warning

Synonyms: Ac-CoA Synthase Inhibitor1

A314446-100MG A314446-10MG A314446-1MG A314446-250MG A314446-25MG A314446-5MG
77167-184EA 452.17 USD
77167-184 77167-186 77167-188 77167-190 77167-192 77167-194
1-(2,3-Di(thiophen-2-yl)quinoxalin-6-yl)-3-(2-methoxyethyl)urea 96%
1-(2,3-Di(thiophen-2-yl)quinoxalin-6-yl)-3-(2-methoxyethyl)urea

Precaución: for Research Use Only

Formula: C₂₀H₁₈N₄O₂S₂
MW: 410.51 g/mol
MDL Number: MFCD03232237
CAS Number: 508186-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR