5-Fluoro-2-methylphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5-Fluoro-2-methylbenzeneboronic acid, 2-Methyl-5-fluorophenylboronic acid

A314220-100G A314220-10G A314220-25G A314220-5G A314220-1G
77194-704EA 120 USD
77194-704 77194-706 77194-708 77194-710 77616-372
5-Fluoro-2-methylphenylboronic acid 98%
5-Fluoro-2-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BFO₂
MDL Number: MFCD03095047
CAS Number: 163517-62-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR