2-(Methylthio)-5-pyridineboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 6-(Methylthio)pyridine-3-boronic acid, 2-Methylthiopyridine-5-boronic acid, 6-(Methylthio)pyridin-3-ylboronic acid

A313924-100MG A313924-1G A313924-250MG
77167-178EA 62.83 USD
77167-178 77167-180 77167-182
2-(Methylthio)-5-pyridineboronic acid 98%
2-(Methylthio)-5-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₆H₈BNO₂S
MW: 169.01 g/mol
MDL Number: MFCD03788238
CAS Number: 321438-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR