5-Methoxy-6-nitrobenzo[d][1,3]dioxole 97%

Supplier: Ambeed
Warning

A313879-100MG A313879-1G A313879-250MG A313879-5G
77194-696EA 9 USD
77194-696 77194-698 77194-700 77194-702
5-Methoxy-6-nitrobenzo[d][1,3]dioxole 97%
5-Methoxy-6-nitrobenzo[d][1,3]dioxole

Precaución: for Research Use Only

Formula: C₈H₇NO₅
MDL Number: MFCD01659655
CAS Number: 10310-02-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR