4-Methyl-1-piperazineacetic acid 97%, Ambeed.Inc

Supplier: Ambeed
Danger

A313243-10G A313243-1G A313243-250MG A313243-25G A313243-5G
77167-158EA 108.15 USD
77167-158 77167-160 77167-162 77683-927 77683-928
4-Methyl-1-piperazineacetic acid 97%, Ambeed.Inc
4-Methyl-1-piperazineacetic acid

Precaución: For research use only.

Formula: C₇H₁₄N₂O₂
MW: 158.1983 g/mol
MDL Number: MFCD03444554
CAS Number: 54699-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR